Find methods for your needs
Separation of L-Cystine, Carboxylic acid and Triethylamine by using UV detection.
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of L-Cystine, Carboxylic acid and Triethylamine. The separation was performed on a Thermo Scientific Hypercarb HPLC column with UV detection at 200 nm. With a total run time of 48 minutes, the method allows easy separation of L-Cystine, Carboxylic acid and Triethylamine.| Market: | Pharma |
| Keywords: | Carboxylic acid, HPLC, UV detection, Hypercarb, L-Cystine, Triethylamine |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.