Find methods for your needs
Fast analysis of artesunate and dihydroartemisinin using a Thermo Scientific Syncronis C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of artesunate and dihydroartemisinin. The separation is performed on a Thermo Scientific Syncronis C18 HPLC column, with UV detection at 210 nm. Considering the the Cmax of artesunate is 0.09+/-0.04µg/mL and that artesunate lacks an intensive chromaphore for UV absorption, MS detection is required to improved sensitivity. Within this application we will demonstrate an alternative method using a MS -compatible mobile phase.| Market: | Pharma |
| Keywords: | Artesunate, Dihydroartemisinin, HPLC, Syncronis C18, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.