Find methods for your needs
CAN120: Determination of Total Petroleum Hydrocarbons in Rubble and Soils by Accelerated Solvent Extraction and GC-FID
Description
Recent advances in Accelerated Solvent Extraction systems, reduces the time spent on sample preparation prior to GC analysis. The method reported here is applicable to soils with a hydrocarbon content (HC)(>C12) between 25 and 4000 mg/kg and waste with a HC (C10-C40) between 50 and 8000 mg/kg expressed on a dry matter basis. All hydrocarbons with a boiling range of approximately 175 °C to 525 °C, e.g. n-alkanes from C10H22 to C40H82, isoalkanes, cycloalkanes, alkyl benzenes, alkyl naphthalenes, and polycyclic aromatic compounds are determined as total petroleum hydrocarbons (TPH).| Market: | Environmental |
| Keywords: | Accelerated solvent extraction, Pressurized Fluid Extraction, Rocket Evaporator, ASE 350, Total Petroleum Hydrocarbons (TPH), Capillary GC column, UNI EN 14039:2005, Hydrocarbons >C12, Flame Ionization Detection, Gas Chromatography, In-Line Clean-Up, Hydrocarbons C10-C40, GC-FID |
| Matrix: | Rubble, Soils |
| Author: | Fabrizio Galbiati,1 Luca Teli, 2 |
| Affiliation: | 1. Thermo Fisher Scientific (Schweiz) AG, Reinach, Switzerland 2. Consulenze Ambientali SpA, Scanzorosciate, Italy |
For Research Use Only. Not for use in diagnostic procedures.