Find methods for your needs
AN356: Determination of Perchlorate in Vegetation Samples Using Accelerated Solvent Extraction and Ion Chromatography
Description
This application note provides the details of using accelerated solvent extraction for the determination of perchlorate in soil, milk, and several plant matrices. The method provides a rapid means of extracting perchlorate from all of the aforementioned matrices using only water as an extraction solvent. The benefits of this method are simplicity, speed of analysis, and automation. Accelerated solvent extraction allows the rapid extraction and in-line cleanup of a large number of samples with minimal labor.| Market: | Food and Beverage |
| Keywords: | ASE, Perchlorate, IonPac AS16 column, ASE 200, ASE 350, Vegetation Samples, IonPac Cryptand C1 Concentrator |
| Matrix: | Vegetation samples |
| Author: | Bruce Richter;Brett Murphy |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.