Find methods for your needs
Improved determination of catecholamines using a Thermo Scientific Syncronis HILIC HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of catecholamines. The separation is performed on a Thermo Scientific Syncronis HILIC HPLC column, with UV detection at 280 nm.| Market: | Pharma |
| Keywords: | 5-hydroxyindole acetic acid, Catechol, Catecholamines, DOPAC, Dopamine, HILIC, HPLC, L-DOPA, Serotonin, Syncronis HILIC, Tyrosine, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.