Find methods for your needs
AU202: A Fast Method for Sugar Analysis of Instant Coffee Samples
Description
The current study updates the fast method described in AN280 with a Dionex CarboPac SA10-4 μm column. Due to the smaller particle size, this column provides more efficient peaks and better resolution, making the sample analysis easier and more reliable. Moreover, the system used for the analysis is updated to the Thermo Scientific™ Dionex™ Integrion™ HPIC™ system. This system combines flexibility and ease-of-use with high sensitivity and selectivity, bringing a new level of convenience and cost effectiveness to simple sugar analysis.| Market: | Food and Beverage |
| Keywords: | Carbohydrates, Electrochemical detection, RFIC, HPAE-PAD, AN280, AOAC Method 995.13, Dionex CarboPac SA10-4μm Column, AU 202, Dionex Integrion HPIC System, TN 20, TN 70671, TN20, TN70671 |
| Matrix: | Water and hydrochloric acid |
| Author: | Sachin Patil and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA |
For Research Use Only. Not for use in diagnostic procedures.