Find methods for your needs
Comparing LC and GC Triple Quadrupole MS for the Screening of 500 Pesticides in Matrix
Description
Modern pesticide analysis is extremely challenging due to the diversity of compounds required to be reported, especially in the area of food safety control. The pressure to report large numbers of pesticides quickly makes it attractive to use large single injection methods. Triple quad MS has emerged as a primary screening technique due to its high sensitivity and selectivity against matrix. Presented is a comparison of both LC and GC sample introduction techniques coupled to triple quad MS screening of over 500 pesticides at ppb levels.| Market: | Environmental |
| Keywords: | Accela UHPLC, TRACE GC, QED, Pesticide Screening, QuEChERS sample prep, TSQ Access MAX MS, TSQ 8000 MS, Data dependent scanning |
| Matrix: | Vegetables |
| Author: | Juan Carmona, David Steiniger, Jason Cole, Paul Silcock, Jonathan Beck, Mary Blackburn, Jennifer Massi, Charles Yang, and Dipankar Ghosh |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.