Find methods for your needs
Measuring Phytosterols in Health Supplements by LC/MS
Description
Plant sterols, phytosterols, are claimed to help lower cholesterol levels in humans. In the year 2000, the United States Food & Drug Administration (US FDA) approved a health claim relating phytosterol ester or phytostanol ester consumption to reduced risk for coronary heart disease. Thus, there is increased interest in qualitative and quantitative analysis of phytosterols in food products. Phytosterols are typically measured by gas chromatography after derivatization. In this study, phytosterols were injected onto an LC-MS for direct analysis.| Market: | Food and Beverage |
| Keywords: | Ultimate UHPLC, MSQ Single QuadMS, Phytosterols, Vitamin supplements |
| Matrix: | Supplements |
| Author: | Marcus Miller and William Schnute |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.