Find methods for your needs
The Thermo Scientific Exactive Benchtop LC-MS Orbitrap Mass Spectrometer
Description
The combination of Orbitrap technology with a linear ion trap has become an established platform for high resolution, accurate mass LC-MSn analysis. The high resolving power, mass accuracy and dynamic range of the Orbitrap analyzer allow rigorous characterization of complex mixtures even in the absence of precursor ion mass selection. We now describe the development of a non-hybrid mass spectrometer comprising of an atmospheric-pressure ion source (API) and a standalone Orbitrap mass analyzer.| Market: | Environmental; Metabolomics; Pharma; BioPharma |
| Keywords: | High resolution, Orbitrap Technology, Exactive, High Mass Accuracy, Polarity switching, Scan speed |
| Matrix: | Various |
| Author: | Andreas Wieghaus, Alexander Makarov, Ulf Froehlich, Markus Kellmann, Eduard Denisov, Oliver Lange |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.