Find methods for your needs
Detection and Quantitation of Brominated and Chlorinated Hydrocarbons by DART with Linear Ion Trap and Triple Quadrupole Technology
Description
Brominated hydrocarbons (BFRs) have been used in various industries for decades. Most chlorinated pesticides (OCs) have been banned in the United States, but their use still occurs in developing countries. The persistence of BFRs and OCs in the environment, potential deleterious activity, makes the detection and monitoring of these compounds an important topic. We propose DART as a simple, rapid, easy-to-use technique; eliminating the need for chromatographic method development, and reducing or eliminating sample preparation, for detection and quantitation of both BFRs and OCs.| Market: | Environmental |
| Keywords: | brominated flame retardants (BFRs), chlorinated pesticides (OCs), DART-SVP source, LTQ Linear ion trap MS, TSQ Quantum Access MAX MS |
| Matrix: | Chlorinated pesticide, Flame retardant |
| Author: | Mary L. Blackburn, Scott J. Harrison, and Maria C. Prieto Conaway |
| Affiliation: | AgResearch, Palmerston North - New Zealand; Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.