Find methods for your needs
A Novel Screening Method for Anthropogenic Sewage Pollutants in Waste Water, Ground Water and Drinking Water Samples by LC-HRAM Analysis
Description
There is a growing public concern about potential contamination of water and the environment with anthropogenic compounds and their degradation products and possible negative impacts on nature and public health. We describe a method that uses the Thermo Scientific Q Exactive mass spectrometer with an Accela Open Autosampler to fully automate the analysis of waste water and drinking water for anthropogenic compounds down to low ppt concentration levels without time consuming solid phase extraction.| Market: | Environmental |
| Keywords: | Water analysis, Accela UHPLC, Q Exactive MS, Anthropogenic pollutants, High Resolution MS (HRMS) |
| Matrix: | Water |
| Author: | Sebastian Westrup, Nick Duczak, and Michal Godula |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.