Find methods for your needs
Simultaneous Analysis of Mycotoxins in Dairy Products by Liquid Chromatography - Quadrupole Orbitrap Mass Spectrometry
Description
Mycotoxins are a diverse group of highly toxic secondary metabolic products from various fungal species. Their analysis can be challenging because the physicochemical properties of these compounds vary widely. LCMS has been widely used, however, this technique is not suitable for simultaneous screening of large numbers of mycotoxins. Instead, ultrahigh-performance LC coupled with hybrid quadrupole-Orbitrap MS can provide both detailed structural information and effective screening of multiple mycotoxins.| Market: | Food and Beverage |
| Keywords: | LC-MS, Data Dependent Analysis, Accela UHPLC System, QuEChERS sample prep, Dairy mycotoxins |
| Matrix: | Milk, Yogurt |
| Author: | Complete method: Jia, W. et al; J. Chrom. A., 2014, 1345, pp 107-114, DOI: 10.1016/j.chroma.2014.04.021 |
| Affiliation: | Shaanxi University of Science & Technology-China; Institute of Food Safety-China |
For Research Use Only. Not for use in diagnostic procedures.