Find methods for your needs
LC-MS/MS Method for the Analysis of Choline and Acetylcholine using a Syncronis HILIC 1.7 µm Column Be the first to rate this application
lcmsms method for analysis choline acetylcholine using a syncronis hilic 17 µm column
# Compound Name Compound Class RT (min)
1 Acetylcholine Neurotransmitter 0.87
2 Choline Neurotransmitter 1.96
Description
Choline is a water soluble essential nutrient that is produced in the body in small quantities. Choline is a precursor of acetylcholine which is an important neurotransmitter involved in muscle control and memory. Choline is a small, highly polar quaternary amine and will not be retained using a C18 column. The zwitterionic nature of Syncronis HILIC makes it ideal for the retention of this class of compound. This application note demonstrates the successful analysis of acetylcholine and choline using a Syncronis HILIC 1.7 µm column.
Market: Clinical Research
Keywords: Acetylcholine, choline, Syncronis HILIC
Matrix: Biological
Author: Eilidh MacRitchie, Ken Meadows
Affiliation: Thermo Fisher Scientific
Uploaded on 3/15/2016.
For Research Use Only. Not for use in diagnostic procedures.