Find methods for your needs
On-line Enrichment HTLC/MS/MS Assay for Multiple Classes of Antibiotics in Environmental Water Sources
Description
There is a growing concern over the presence of antibiotics in environmental sources of water. This has caused environmental and government labs to develop LC/MS methods to monitor water supplies for the presence of antibiotics. We developed a method to screen for antibiotics in surface water by applying on-line preconcentration and analyte extraction with LC/MS/MS detection.| Market: | Environmental |
| Keywords: | TSQ Quantum Ultra, Aria TLX-2, H-SRM, Turbulent-flow Extraction |
| Matrix: | Water |
| Author: | Kevin J. McHale, Chris Esposito, and Francois Espourteille |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.