Find methods for your needs
LC-MS/MS Analysis of Herbicides in Drinking Water at Femtogram Levels Using 20 mL EQuan Direct Injection Techniques
Description
LC-MS/MS is routinely used by the environmental and food industries to identify and quantify pesticide and herbicide residues. However, this method typically requires extensive offline sample preconcentration methods, which can be expensive and time-consuming, to meet the stringent requirements and low limits of detection set forth by federal and international regulatory authorities. An online preconcentration and cleanup method has been developed that improves both sensitivity and precision and yields unmatched throughput.| Market: | Environmental |
| Keywords: | Herbicides, Water analysis, QED |
| Matrix: | Water |
| Author: | Jonathan R. Beck, Charles Yang |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.