Find methods for your needs
AN533: Analysis of Perchlorate in Infant Formula by Ion Chromatography-Electrospray-Tandem Mass Spectrometry (IC-ESI-MS/MS)
Description
A simple and ultra-sensitive IC-MS/MS technique for the quantitation of perchlorate in powdered infant formula, liquid infant formula, and milk is described here. Unlike conventional detection methods that require labor intensive solid phase extraction (SPE) sample enrichment, only a simple sample preparation is necessary.| Market: | Food and Beverage |
| Keywords: | ESI, sensitivity, ion chromatography, Food Safety, IC MS/MS, AN63436 |
| Matrix: | Infant formula, Milk |
| Author: | Charles Yang, Leo Wang, and William Schnute |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.