Find methods for your needs
Determination of Inorganic Cations and Ammonium in Environmental Waters by Ion Chromatography Using the Dionex IonPac CS16 Column with a Compact Ion Chromatography System
Description
This record describes the use of IC with a 5 mm i.d. Dionex IonPac CS16 cation-exchange column, an electrolytically generated methanesulfonic acid (MSA) eluent, and suppressed conductivity detection to determine dissolved alkali and alkaline earth cations and ammonium in municipal wastewater as published in Application Note 141. In this record the method was executed on an Integrion ion chromatography system.| Market: | Environmental |
| Keywords: | IonPac CS16, Effluent, RFIC, ion chromatography, IC, wastewater, cations, Discharge, Integrion RFIC, RFIC System, cation-exchange chromatography |
| Matrix: | Municipal wastewater |
| Author: | Carl Fisher |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.