Find methods for your needs
Determination of Ammonia in Sodium Bicarbonate Using a Compact Ion Chromatography System
Description
One of the main pharmaceutical uses of sodium bicarbonate is for hemodialysis. For hemodialysis-grade sodium bicarbonate, the quality and purity requirements are stringent. Ammonia is one of the impurities that is assayed in sodium bicarbonate because the presence of elevated levels of ammonia in the blood stream can have adverse effects. This record describes an ion chromatography (IC)-based method for the determination of ammonia (as ammonium) in sodium bicarbonate that was published in Application Note 1073. In this record the method was executed using an Integrion IC system.| Market: | Pharma |
| Keywords: | IonPac CS16, Hemodialysis Solution, RFIC, USP-NF, Sodium hydrogen carbonate |
| Matrix: | Sodium bicarbonate solution |
| Author: | Carl Fisher |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.