Find methods for your needs
Anion Determinations on 4 mm IonPac AS25 Column for Sulfur Speciation
Description
Anion Determinations on 4 mm IonPac AS25 and IonPac AG 20 Guard Column for Sulfur Speciation. This column is optimized for sulfur speciation and resolution of sulfate from carbonate. This column is ideal for sulfite determinations in environmental samples.| Market: | Environmental |
| Matrix: | Water |
For Research Use Only. Not for use in diagnostic procedures.