Find methods for your needs
Related substances analysis of Pregabalin on solid dosages
Description
The Thermo Scientific™ Dionex™ UltiMate 3000 LC system is applied for a related substance analysis of pregabalin. The separation was performed on a Thermo Scientific Accucore PFP 2.6 µm HPLC column with UV detection.| Market: | Pharma |
| Keywords: | Accucore PFP, Pregabalin, UltiMate 3000 RS |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.