Find methods for your needs
TN46: Determination of Trace Anions in Concentrated Glycolic Acid
Description
This Technical Note describes the theory, set up, and analytical procedure for the determination of trace chloride and sulfate at sub-mg/L (ppm) levels in 0.7-17.5% (v/v) glycolic acid.| Market: | Material Science |
| Keywords: | Glycolate, IonPac AS9-HC, IonPac ICE-AS6, TN 46, IonPac AG9-HC |
| Matrix: | Glycolic acid |
For Research Use Only. Not for use in diagnostic procedures.