Find methods for your needs
AB108: Determination of Phosphite in Electroless Nickel Plating Bath Be the first to rate this application
phosphite electroless nickel plating bath
No compound information available.
Description
Electroless nickel plating relies on a reducing agent, such as sodium hypophosphite, to deposit nickel phosphorous on the surface of an item. Over time, the hypophosphite in the bath will oxidize to phosphite. This Application Brief uses an IonPac AS17 column and a Regent-Free IC system to determine phosphite in an electroless nickel plating bath.
Market: Other
Keywords: Chloride, phosphate, IonPac AS17-C, Sulfate, nickel, phosphite, Plating bath, RFIC, Electroless
Matrix: Plating bath
Author: Weerapong Worawirunwong and Jeffrey Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 10/29/2015.
For Research Use Only. Not for use in diagnostic procedures.