Find methods for your needs
TN42: Glycoprotein Oligosaccharide Analysis Using High-Performance Anion-Exchange Chromatography
Description
In this Technical Note, accuracy, precision, linearity, and limits of detection are documented for a commercially available mix of bovine fetuin N-linked oligosaccharide alditols using an HPAE-PAD method. These results are intended as a guide for investigators who need to validate the performance of HPAE-PAD for oligosaccharide mapping. Readers are also encouraged to see newer AppsLab record for this analysis including AN1050, AU176, and AN215.| Market: | BioPharma |
| Keywords: | carbohydrate, Electrochemical detection, glycoprotein, oligosaccharide, PAD, Bovine fetuin, AU 176, HPAE-PAD, TN21, TN71, TN 20, TN 70671, TN20, TN70671, TN 42, oligosaccharide mapping |
| Matrix: | Glycoprotein |
| Author: | Michael Weitzhandler |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.