Find methods for your needs
AU138: Determination of Ethanolamines in Industrial Waters by Cation-Exchange Chromatography
Description
Ion chromatography has been successfully used to determine amines in a variety of chemical, petrochemical, and environmental samples. However, analysis of some samples can be hampered by large concentrations of one or more amines, making the determination of trace concentrations of other amines and cations difficult. The selectivity of the IonPac CS11 column with an methanesulfonic acid eluent addresses many of these challenging analyses. This AU describes the setup and performance of this method. In this record we provide the consumables to update this method with eluent generation.| Market: | Chemical |
| Keywords: | Diethanolamine, methanesulfonic acid, triethanolamine, suppressed conductivity detection, IonPac CS11 column, industrial water, ethanolamines, cation exchange chromatography, Methyldiethanolamine, N-Methyldiethanolamine |
| Matrix: | Industrial waters |
| Author: | Edward Kaiser and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.