Find methods for your needs
AB165: Determination of p-Toluenesulfonic Acid in Water-Insoluble Drugs Be the first to rate this application
ab165 determination ptoluenesulfonic acid waterinsoluble drugs
Description
Here, the convenience and accuracy of a Reagent-Free IC (RFIC) approach is applied to the determination of p-toluenesulfonic acid in water-insoluble drugs using an on-line matrix-elimination as described in Application Note 220, albeit with different columns and a different system configuration. (Note- The method discussed here requires Thermo Scientific Dionex AS-AP Autosampler with optional valves (one 6-port valve and one 10-port valve for sequential injection).
Market: Pharma
Keywords: API, Pharmaceutical, AB 165, AN 220, IonPac AS22, IonPac NG1 Guard, RFIC, Organic Acid Analysis, Tosylate, insoluble API, insoluble drug, AB165, AN220
Matrix: Methanol / water
Author: Pan Guangwen, Liao Dongli, Xu Qun, Jeffrey Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 10/1/2015.
For Research Use Only. Not for use in diagnostic procedures.