Find methods for your needs
Determination of Common Inorganic Cations in Drinking Water Using the IonPac CS16 Column QAR Method
Description
A simple ion chromatography method for the direct determination of inorganic cations in drinking water by Reagent Free ICS 5000 plus system, requiring only deionized water to electrolytically produce a MSA eluent. The method in this record is the Quality Assurance Report (QAR) method, and therefore can be used to test the performance of a Dionex IonPac CS16 column.| Market: | Environmental |
| Keywords: | Drinking Water, Ion Chromatography (IC), IonPac CS16 column, Suppressed Conductivity, CERS 500 |
| Matrix: | Drinking water |
| Author: | Jingli Hu; Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.