Find methods for your needs
Determination of Chlorine and Sulfur in Wood Chips Using Combustion Ion Chromatography
Description
Wood chips are used for producing paper and are also contained in waste deposits. It is important to know the halogen contents of wood chips for impurity management in paper production and for environmental assessment in waste materials. By using a combustion ion chromatography (CIC) system consisting of the automatic quick furnace AQF-2100H and an ICS-1600 ion chromatograph, the concentrations of halogens and sulfur can be determined accurately and quickly.| Market: | Environmental |
| Keywords: | power plant, Combustion, IonPac AS12A, Sulfur, Halogen, boiler, Municipal waste, Chlorine, Fluorine, Corrosive, environmental contaminant, AQF-2100H, TN000767 |
| Matrix: | Wood chips |
For Research Use Only. Not for use in diagnostic procedures.