Find methods for your needs
EP 8.0 monograph - impurity determination of ceftriaxone sodium using a C18 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of ceftriaxone sodium and its impurities. The separation was performed on a Thermo Scientific Syncronis C18 HPLC column using the method described in the EP 8.0 monograph. The results obtained for resolution exceeded the criteria stated in the EP monograph. Resolution between ceftriaxone sodium and impurity A (ceftriaxone sodium E isomer) was 10.0 (EP criteria of minimum 3.0).| Market: | Pharma |
| Keywords: | HPLC, Syncronis C18, UV detection, EP 8.0 monograph, Ceftriaxone sodium |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.