Find methods for your needs
AU146: Determination of Anions in Acid Rain by Ion Chromatography
Description
In this application update, we describe the use of automated eluent generation combined with a high-capacity, hydroxide-selective, anion-exchange column for the determination of inorganic anions in rainwater. This document updates Application Note 31 by incorporating electrolytic eluent generation with the Dionex IonPac AS18 column to eliminate the need for manually preparing the eluents off- line, and thereby increasing the IC system’s level of automation and ease of use.| Market: | Environmental |
| Keywords: | Chloride, fluoride, IonPac AS18, Phosphate, Bromide, Nitrate, RFIC, Sulfate, Acid rain |
| Matrix: | Water |
| Author: | Brian De Borba and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.