Find methods for your needs
Ternary Gradient for Tenofovir Disoproxil Fumarate Impurity Profiling Be the first to rate this application
1
2
3
4
5
6
7
8
9
0.511.522.533.544.555.5Time [min]-1000100200300400500600700800Response [mAU]
# Compound Name Compound Class RT (min)
1 Adenine Related Compound 0.75
2 Tenofovir Related Compound 1.10
3 Emtricitabine Active pharmaceutical ingredient 1.92
4 Emtricitabine Impurity Impurity 2.70
5 TDF impurity 1 Impurity 3.15
6 TDF impurity 2 Impurity 3.88
7 TDF impurity 3 Impurity 3.90
8 Tenofovir Disoproxil Fumarate (TDF) Active pharmaceutical ingredient 4.46
9 TDF impurity 4 Impurity 4.73
Description
The antiretroviral drug tenofovir is in formulation given as the prodrug tenofovir disoproxil fumarate (TDF) in combination with the nucleoside reverse-transcriptase inhibitor emtricitabine. Here we show the impurity profiling of an active pharmaceutical ingredient with the Thermo Scientfic Vanquish Flex UHPLC system. The simultaneous separation of early-eluting polar compounds and later-eluting nonpolar compounds is achieved by applying a ternary gradient with the quaternary Vanquish F pump using a Thermo Scientific Accucore aQ column.
Market: Pharma
Keywords: Accucore aQ, Active pharmaceutical ingredients, USP, aqueous gradient, zero percent organic
Matrix: N/A
Author: Susanne Fabel
Affiliation: Thermo Fisher Scientific
Uploaded on 6/17/2015.
For Research Use Only. Not for use in diagnostic procedures.