Find methods for your needs
AU139: Determination of an Anionic Fluorochemical Surfactant (FC-95) in a Steel Bath Be the first to rate this application
au139 determination an anionic fluorochemical surfactant fc95 a steel bath
# Compound Name Compound Class RT (min)
1 FC-95 Fluorochemical Surfactant 7.50
Description
This Application Update describes a method for determining low ppm amounts of FC-95 in a steel bath (concentrated hydrofluoric, hydrochloric, and nitric acids with 200 g/L each of iron, chromium, and nickel) by ion chromatography. The method described in this Update is similar to that described in AN 119. The surfactant is removed from the acidic matrix and concentrated on an IonPac® NG1 column. The concentrated surfactant is then eluted from the IonPac NG1 column onto an OmniPac PAX-500 column set and detected by suppressed conductivity.
Market: Chemical
Keywords: IC, ion chromatography, ionic surfactant, IonPac NG1, matrix elimination, OmniPac PAX-500 column, online sample preparation, suppressed conductivity
Matrix: Steel bath
Author: Mark Laikhtman and Jeff Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 6/5/2015.
For Research Use Only. Not for use in diagnostic procedures.