Find methods for your needs
Characterization of of Pluronic F127 Surfactant by HPLC-CAD Be the first to rate this application
Description
Pluronic F127, or Poloxamer 407, is a non-ionic, polyol surfactant. Due to the variety of different structures contained within this complex product, a characterization method is described to determine the relative amounts of the different components, which may vary from lot-to-lot.
Market: BioPharma
Keywords: Poloxamer 407, Pluronic F127
Matrix: Water
Uploaded on 5/28/2015.
For Research Use Only. Not for use in diagnostic procedures.