Find methods for your needs
Analysis of phenols and chlorinated phenols in drinking water by GC/MS using EPA method 528
Description
The Thermo Scientific TRACE GC Ultra is applied for the analysis of phenols and chlorinated phenols in drinking water by EPA method 528. The separation was performed on a Thermo Scientific TraceGOLD TG-5SilMS GC column, with MS detection.| Market: | Environmental |
| Keywords: | Chlorinated phenols, Drinking water, EPA 528, GC, MS detection, Phenols, TraceGOLD TG-5SilMS |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.