Find methods for your needs
AN85: Determination of Trace Anions in (Acetone) Organic Solvents
Description
The determination of trace anionic contamination in processing solvents (N-methyl-2-pyrrolidone, isopropyl alcohol, and acetone) is important to the electronics industries to minimize corrosion. Here matrix elimination is used to retain and concentrate µg/L anionic contamination in a 5 mL acetone sample onto a concentrator column. Acetone is flushed to waste (5 min) with purified deionized water (10 mL). Trace anions are eluted from the concentrator and separated on IonPac AS9-HC column using 8 mM carbonate, 1.5 mM NaOH eluent. AU163 updates AN85 with < 0.2 ug/L lower detection limits.| Market: | Chemical |
| Keywords: | Acetone, Nitrate, chloride, electronic grade solvent, electronics, IonPac AS9-HC column, matrix elimination, sulfate, trace anions, semiconductor grade solvent |
| Matrix: | Acetone |
For Research Use Only. Not for use in diagnostic procedures.