Find methods for your needs
Simple determination of sorbitol using HPLC with RI detection
Description
In this work the Thermo Scientific Hypersil GOLD Amino column was used for the determination of sorbitol. A simple isocratic method was used with RI detection and a run time of seven minutes was achieved.| Market: | Food and Beverage |
| Keywords: | HPLC, Hypersil GOLD Amino, RI detection, Sorbitol, Sugar alcohol |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.