Description
The Thermo Scientific HPLC system is applied for the analysis of phenol standards by EPA method 604. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage (PA) column, with UV detection at 285 nm.| Market: | Chemical |
| Keywords: | Acclaim PolarAdvantage, Chlorophenol, Dichlorophenol, Dimethylphenol, Dinitrophenol, EPA 604, HPLC, Nitrophenol, Pentachlorophenol, Phenol, Trichlorophenol, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.