Description
The Thermo Scientific HPLC system is applied for the analysis of glycolic and related acids. The separation is performed on a Thermo Scientific Acclaim Organic Acid (OA) column, with UV detection at 210 nm.| Market: | Chemical |
| Keywords: | Acclaim Organic Acid, Acetic acid, Formic acid, Glycolate, HPLC, Lactic acid, Organic acid, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.