Find methods for your needs
Simple isocratic method for the separation of fat-soluble vitamins
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of fat-soluble vitamins. The separation is performed using a Thermo Scientific Acclaim Phenyl-1 column, with UV detection at 220 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim Phenyl-1, Fat-soluble vitamins, HPLC, Tocopherol, trans-Retinol acetate, UV detection, Vitamin D2, Vitamin D3, Vitamin E, Vitamin K1, Vitamin K2 |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.