Find methods for your needs
High resolution characterization of poly-2-methyl-2-oxazoline by HPLC-CAD
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the characterization of poly-2-methyl-2-oxazoline. The separation is performed on a Thermo Scientific Acclaim C30 HPLC column, with charged aerosol detection (CAD).| Market: | Material Science |
| Keywords: | Acclaim C30, CAD, Charged aerosol detection, HPLC, Poly-2-methyl-2-oxazoline, Polymer, Toluene sulfonate |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.