Find methods for your needs
Quick analysis of polyacrylic acid with comparison of CAD and UV detection
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the size exclusion chromatography analysis of polyacrylic acid. The separation is performed on a Thermo Scientific Acclaim SEC-1000 HPLC column, with both charged aerosol detection (CAD) and UV detection. The CAD provides superior results with LOQ around 0.015 µg/mL.| Market: | Environmental |
| Keywords: | Polymer, Acclaim SEC-1000, CAD, Charged aerosol detection, HPLC, PAA, Polyacrylic acid, SEC, Size exclusion chromatography, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.