Find methods for your needs
Accurate determination of atorvastatin impurities using reversed-phase HPLC-UV
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of atorvastatin impurities. The separation was performed on a Thermo Scientific Accucore aQ HPLC column using a reversed-phase method with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | Accucore aQ, Atorvastatin, HPLC, UV detection, Impurities |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.