Find methods for your needs
AN31: Determination of Anions in Acid Rain by Ion Chromatography
Description
This application note describes the established ion chromatographic method for the determination of anions in rainwater samples extended to the use of microbore columns and accessories. A rainwater standard from the National Institute of Standards and Technology (NIST) was analyzed, verifying the accuracy of the method. This method is updated in AU146 to include the use of electrolytic eluent generation and the IonPac AS18 column.| Market: | Environmental |
| Keywords: | Bromide, Chloride, Nitrate, Nitrite, Phosphate, Sulfate, Fluoride, Acid rain, NIST, SRM 2694-ll, Pollution, IonPac AS4A-SC |
| Matrix: | Water |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.