Find methods for your needs
Accurate assay analysis of fenpiverinium, pitofenone and diclofenac in parental formulation by HPLC-UV
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of fenpiverinium, pitofenone and dicofenac in formulation. The separation was performed on a Thermo Scientific Accucore C18 HPLC column with UV detection at 210 and 286 nm.| Market: | Pharma |
| Keywords: | Accucore C18, Diclofenac, Formulation, HPLC, UV detection, Fenpiverinium, Pitofenone |
| Matrix: | Formulation |
For Research Use Only. Not for use in diagnostic procedures.