Find methods for your needs
Improved analysis of laureth sulfate and coco-amidopropyl betaine in shampoos by HPLC-CAD
improved analysis laureth sulfate cocoamidopropyl betaine shampoos by hplccad
Description
The Thermo Scientific UltiMate 3000 LC system is applied for the analysis of laureth sulfate and coco-amidopropyl betaine in shampoos. The separation is performed on a Thermo Scientific Acclaim Surfactant Plus column, with charged aerosol detection.
Market: Other
Keywords: Acclaim Surfactant Plus, CAD, Charged aerosol detection, HPLC, Laureth sulfate, Shampoo, Surfactant, Coco-amidopropyl betaine
Matrix: Shampoo
Uploaded on 3/5/2015.
For Research Use Only. Not for use in diagnostic procedures.