Find methods for your needs
Detailed profiles of two laundry detergents by HPLC-CAD Be the first to rate this application
detailed profiles two laundry detergents by hplccad
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of two laundry detergents. The separation is performed on a Thermo Scientific Acclaim Surfactant Plus column, with charged aerosol detection.
Market: Other
Keywords: Surfactant, Acclaim Surfactant Plus, CAD, Charged aerosol detection, HPLC, Laundry detergent, Dodecyl benzene sulfonate, LAS
Matrix: Laundry detergent
Uploaded on 3/5/2015.
For Research Use Only. Not for use in diagnostic procedures.