Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of two laundry detergents. The separation is performed on a Thermo Scientific Acclaim Surfactant Plus column, with charged aerosol detection.| Market: | Other |
| Keywords: | Surfactant, Acclaim Surfactant Plus, CAD, Charged aerosol detection, HPLC, Laundry detergent, Dodecyl benzene sulfonate, LAS |
| Matrix: | Laundry detergent |
For Research Use Only. Not for use in diagnostic procedures.