Find methods for your needs
Improved analysis of anionic and nonionic surfactants in dishwasher rinsing aid using a Thermo Scientific Acclaim Surfactant Plus column with charged aerosol detection
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of anionic and nonionic surfactants in dishwasher rinsing aid. The separation is performed on a Thermo Scientific Acclaim Surfactant Plus column, with charged aerosol detection.| Market: | Environmental |
| Keywords: | Acclaim Surfactant Plus, Anionic, CAD, Charged aerosol detection, Cumene sulfonate, HPLC, Nonionic, Rinsing aid, Surfactants |
| Matrix: | Rinsing aid |
For Research Use Only. Not for use in diagnostic procedures.