Find methods for your needs
AN297: Determination of Total and Potential Sulfate and Total Chloride in Fuel-Grade Butanol Per ASTM D7319-09
Description
Here we present a direct injection IC approach to determine total and potential sulfate and total chloride in butanol intended as a gasoline additive. This IC method is consistent with the procedure described in ASTM D7319-09, a standard intended for the analysis of ethanol samples containing 1.0–20 mg/kg of total or potential inorganic sulfate and 1.0–50 mg/kg of inorganic chloride. This method also has the sensitivity to meet the specifications described in ASTM D4806-11A.| Market: | Petrochem, Oil and Gas |
| Keywords: | biofuels, Chloride, Sulfate, ASTM D4806-11A, High Octane Rating, Renewable Energy |
| Matrix: | Butanol |
| Author: | Pranathi Perati, Brian DeBorba, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.