Find methods for your needs
AU185: Determination of Nitrite and Nitrate in Wastewater Using Capillary IC with UV Detection
Description
Ion chromatography (IC) with suppressed conductivity detection is an effective technique to simultaneously determine common inorganic anions in water samples. However determining nitrate and nitrite at low concentrations in high ionic strength samples such as mineral water, wastewater, and brine, can be challenging. IC with UV detection provides an alternate approach for determining ions with chromophore properties without compromising sensitivity. This method updates AU132 using a Dionex IonPac AS18-Fast capillary column on a capillary IC system. An ICS-6000 can be used for this application.| Market: | Environmental |
| Keywords: | ICS-5000+, HPIC, IonPac AS18-Fast, waste water, Nitrate, UV detection, RFIC, Nitrite, IonPac AS18, capillary IC, ICS-5000, wastewater, ICS-6000 |
| Matrix: | Municipal wastewater |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.