Find methods for your needs
Fast analysis of five antidepressants using a Thermo Scientific Acclaim PolarAdvantage (PA) HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of five antidepressants. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage (PA) HPLC column, with UV detection at 220 nm.| Market: | Pharma |
| Keywords: | Acclaim PolarAdvantage (PA), Amitriptyline, Antidepressants, Doxepin, HPLC, Imipramine, Nortriptyline, Protriptyline, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.